Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMA3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005914.2 | NP_001005914.1 | 18 | 2958 | ||
NM_001290060.1 | NP_001276989.1 | 17 | 2915 | |||
NM_001290061.1 | NP_001276990.1 | 17 | 2764 | |||
NM_001290062.1 | NP_001276991.1 | 11 | 2181 | |||
NM_001290063.1 | NP_001276992.1 | 11 | 2055 | |||
NM_004636.3 | NP_004627.1 | 18 | 2961 | |||
NR_110697.1 | 8 | 1682 | ||||
GenBank mRNA | AB209322.1 | - | 16 | 3141 | ||
AF217991.1 | - | 17 | 2724 | |||
AI309508.1 | - | 1 | 203 | |||
AK092182.1 | - | 8 | 1644 | |||
AK098329.1 | - | 5 | 3247 | |||
BC009113.1 | - | 14 | 2665 | 4153377 | ||
BC013975.2 | - | 17 | 2738 | 4121913 | ||
BC024220.2 | - | 17 | 2738 | 3009807 | ||
BC044602.1 | - | 11 | 2143 | |||
U28369.1 | - | 17 | 2904 |