Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMA3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005914.2 | NP_001005914.1 | 12 | 1564 | ||
NM_001290060.1 | NP_001276989.1 | 11 | 1521 | |||
NM_001290061.1 | NP_001276990.1 | 11 | 1370 | |||
NM_001290062.1 | NP_001276991.1 | 5 | 787 | |||
NM_001290063.1 | NP_001276992.1 | 5 | 661 | |||
NM_004636.3 | NP_004627.1 | 12 | 1567 | |||
NR_110697.1 | 3 | 446 | ||||
GenBank mRNA | AB083186.1 | - | 11 | 1317 | ||
AB209322.1 | - | 10 | 1747 | |||
AF217991.1 | - | 11 | 1330 | |||
AK092182.1 | - | 3 | 408 | |||
AK098329.1 | - | 2 | 1071 | |||
AK293678.1 | - | 5 | 620 | |||
BC009113.1 | - | 8 | 1271 | 4153377 | ||
BC013975.2 | - | 11 | 1344 | 4121913 | ||
BC024220.2 | - | 11 | 1344 | 3009807 | ||
BC044602.1 | - | 5 | 749 | |||
U28369.1 | - | 11 | 1510 |