Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRINA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000837.1 | NP_000828.1 | 7 | 1563 | ||
NM_001009184.1 | NP_001009184.1 | 7 | 1451 | |||
GenBank mRNA | AB097026.1 | - | 7 | 1375 | ||
AK127640.1 | - | 7 | 1599 | |||
AK294127.1 | - | 8 | 1308 | |||
AL157442.1 | - | 1 | 294 | |||
BC041788.1 | - | 7 | 1409 | 4738372 | ||
BC065834.1 | - | 6 | 1317 | |||
BC084553.1 | - | 6 | 1300 | 6206624 |