Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016298.3 | NP_057382.2 | 3 | 614 | ||
GenBank mRNA | AB033021.1 | - | 3 | 818 | ||
AF204674.1 | - | 3 | 520 | |||
AK314400.1 | - | 3 | 536 | |||
BC109275.1 | - | 2 | 425 | 40028315 | ||
BC109276.1 | - | 2 | 425 | 40028318 |