Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP13A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020410.2 | NP_065143.2 | 25 | 3446 | ||
GenBank mRNA | AB058728.1 | - | 25 | 3416 | ||
AF288687.1 | - | 25 | 3064 | |||
AK026044.1 | - | 9 | 1596 | |||
AK056420.1 | - | 25 | 3422 | |||
AK074159.1 | - | 15 | 4314 | |||
AK074798.1 | - | 13 | 1951 | |||
AK074920.1 | - | 14 | 2026 | |||
AK095287.1 | - | 8 | 2619 | |||
AK123780.1 | - | 7 | 1873 | |||
AK125861.1 | - | 8 | 3826 | |||
AK130125.1 | - | 16 | 2086 | |||
AK172778.1 | - | 24 | 2877 | |||
AL834180.1 | - | 2 | 1895 | |||
BC009302.2 | - | 7 | 890 | |||
BC069211.1 | - | 6 | 780 |