Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TECPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172631.2 | NP_001166102.1 | 13 | 3336 | ||
NM_014844.4 | NP_055659.2 | 13 | 3336 | |||
GenBank mRNA | AB002295.1 | - | 10 | 2639 | ||
AB002327.1 | - | 13 | 3285 | |||
AK299690.1 | - | 6 | 736 | |||
BC136647.1 | - | 12 | 3133 | 9020637 | ||
BC142667.1 | - | 13 | 3169 | 40148157 | ||
BC142715.1 | - | 13 | 3169 | 40148843 | ||
BC151230.1 | - | 13 | 3309 | 8860116 |