Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004041.4 | NP_004032.2 | 2 | 264 | ||
NM_020251.3 | NP_064647.1 | 2 | 264 | |||
XM_011545034.2 | XP_011543336.1 | 3 | 148 | |||
XM_011545035.2 | XP_011543337.1 | 3 | 148 | |||
XM_017017750.1 | XP_016873239.1 | 3 | 148 | |||
XM_017017751.1 | XP_016873240.1 | 3 | 148 | |||
XM_017017752.1 | XP_016873241.1 | 2 | 104 | |||
XM_017017753.1 | XP_016873242.1 | 2 | 168 | |||
XM_017017754.1 | XP_016873243.1 | 2 | 104 | |||
GenBank mRNA | AF084040.1 | - | 2 | 40 | ||
AF084940.1 | - | 2 | 40 | |||
AK289718.1 | - | 2 | 94 | |||
BC003636.2 | - | 2 | 117 | 3604829 | ||
BQ883950.1 | - | 2 | 287 | |||
FJ348262.1 | - | 2 | 40 | |||
L04685.1 | - | 2 | 40 |