Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001844.4 | NP_001835.3 | 54 | 4529 | ||
NM_033150.2 | NP_149162.2 | 53 | 4322 | |||
XM_017018828.1 | XP_016874317.1 | 56 | 4614 | |||
XM_017018829.1 | XP_016874318.1 | 56 | 4611 | |||
XM_017018830.1 | XP_016874319.1 | 55 | 4404 | |||
XM_017018831.1 | XP_016874320.1 | 54 | 5914 | |||
GenBank mRNA | AL834148.1 | - | 37 | 3434 | ||
BC007252.1 | - | 7 | 1024 | |||
BC116449.1 | - | 54 | 4350 | 7486698 | ||
BT007205.1 | - | 4 | 691 | |||
X06268.1 | - | 5 | 826 | |||
X16468.1 | - | 53 | 4141 |