Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATPIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016311.4 | NP_057395.1 | 2 | 257 | ||
NM_178190.2 | NP_835497.1 | 2 | 257 | |||
NM_178191.2 | NP_835498.1 | 2 | 257 | |||
GenBank mRNA | AB029042.1 | - | 2 | 185 | ||
AF114836.1 | - | 343 | ||||
AK000934.1 | - | 2 | 194 | |||
AK316600.1 | - | 2 | 244 | |||
AL050386.1 | - | 2 | 187 | |||
AL583540.3 | - | 2 | 180 | |||
BC004955.1 | - | 2 | 140 | 3625379 | ||
BC009677.1 | - | 2 | 215 | 3877506 | ||
BG036978.1 | - | 2 | 229 | |||
BG337531.1 | - | 2 | 213 | |||
BG714388.1 | - | 2 | 250 | |||
BP271519.1 | - | 2 | 253 | |||
BT009849.1 | - | 2 | 164 | |||
CR457097.1 | - | 2 | 164 | |||
DB030607.1 | - | 2 | 257 |