Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEKR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004316.2 | NP_001004316.2 | 13 | 1959 | ||
XM_005247441.3 | XP_005247498.1 | 13 | 3771 | |||
XM_006713631.3 | XP_006713694.1 | 13 | 2483 | |||
XM_006713636.2 | XP_006713699.1 | 8 | 1179 | |||
XM_011512803.2 | XP_011511105.1 | 12 | 2183 | |||
XM_011512804.2 | XP_011511106.1 | 13 | 4004 | |||
XM_011512809.1 | XP_011511111.1 | 10 | 1590 | |||
XM_011512813.2 | XP_011511115.1 | 8 | 1263 | |||
XM_011512814.2 | XP_011511116.1 | 8 | 1171 | |||
XM_017006374.1 | XP_016861863.1 | 9 | 1367 | |||
XM_017006375.1 | XP_016861864.1 | 9 | 1358 | |||
GenBank mRNA | AK131473.1 | - | 14 | 2268 |