Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR1L8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004454.1 | NP_001004454.1 | 1 | 730 | ||
XM_017014284.1 | XP_016869773.1 | 3 | 1266 | |||
XM_017014285.1 | XP_016869774.1 | 3 | 1266 | |||
XM_017014286.1 | XP_016869775.1 | 2 | 1137 | |||
GenBank mRNA | BC140910.1 | - | 1 | 812 | 9056722 |