Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABI2 SIRNA Assays ›
Gene Symbol:
ABI2
Gene Name:
abl-interactor 2
Gene Aliases:
ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB
Chromosome Location:
Chr.2: 203327874 - 203432173 on Build GRCh38
UniGene ID:
Hs.471156
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
241470
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABI2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282925.1
NP_001269854.1
9
1447
NM_001282926.1
NP_001269855.1
8
1429
NM_001282927.1
NP_001269856.1
7
922
NM_001282932.1
NP_001269861.1
8
1443
NM_005759.5
NP_005750.4
7
1246
XM_005246217.2
XP_005246274.1
9
1812
XM_005246218.2
XP_005246275.1
8
1794
XM_005246221.2
XP_005246278.1
7
1647
XM_005246225.2
XP_005246282.1
7
1647
XM_005246226.2
XP_005246283.1
7
1611
XM_006712174.2
XP_006712237.1
9
1812
XM_006712177.2
XP_006712240.1
10
1888
XM_006712179.2
XP_006712242.1
8
1665
XM_006712180.2
XP_006712243.1
8
1629
XM_006712181.2
XP_006712244.1
8
1629
XM_006712182.2
XP_006712245.1
8
1626
XM_006712185.1
XP_006712248.1
8
1687
XM_006712186.2
XP_006712249.1
6
1464
XM_011510472.2
XP_011508774.1
8
1662
XM_011510473.2
XP_011508775.1
8
1111
XM_017003118.1
XP_016858607.1
9
1809
XM_017003119.1
XP_016858608.1
9
1812
XM_017003120.1
XP_016858609.1
9
1809
XM_017003121.1
XP_016858610.1
9
1809
XM_017003122.1
XP_016858611.1
8
1665
XM_017003123.1
XP_016858612.1
7
1644
XM_017003124.1
XP_016858613.1
7
1611
XM_017003125.1
XP_016858614.1
7
1608
XM_017003126.1
XP_016858615.1
7
1611
XM_017003127.1
XP_016858616.1
7
1608
XM_017003128.1
XP_016858617.1
10
1888
XM_017003129.1
XP_016858618.1
8
1665
XM_017003130.1
XP_016858619.1
8
1662
XM_017003131.1
XP_016858620.1
7
1644
XM_017003132.1
XP_016858621.1
8
1629
XM_017003133.1
XP_016858622.1
9
1741
XM_017003134.1
XP_016858623.1
7
1611
XM_017003135.1
XP_016858624.1
7
1608
XM_017003136.1
XP_016858625.1
7
1608
XM_017003137.1
XP_016858626.1
9
1702
XM_017003138.1
XP_016858627.1
7
1479
XM_017003139.1
XP_016858628.1
8
1687
XM_017003140.1
XP_016858629.1
8
1684
XM_017003141.1
XP_016858630.1
6
1464
XM_017003142.1
XP_016858631.1
6
1461
XM_017003143.1
XP_016858632.1
9
1738
XM_017003144.1
XP_016858633.1
7
1482
XM_017003145.1
XP_016858634.1
6
1461
XM_017003146.1
XP_016858635.1
6
1464
GenBank mRNA
AF260261.1
-
9
1171
AK294429.1
-
3
499
AK297537.1
-
4
388
AK299092.1
-
6
584
AK299672.1
-
7
737
AK299679.1
-
3
365
AK299688.1
-
4
538
AK299750.1
-
2
184
AK299824.1
-
8
1273
AK302853.1
-
8
1369
AK316343.1
-
2
238
BC001439.2
-
7
995
3138341
BT009920.1
-
7
970
U23435.1
-
7
883
U31089.1
-
6
715
X95632.1
-
7
1070
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
movement of cell or subcellular component
cytoskeleton organization
actin polymerization or depolymerization
viral process
cell migration
Rac protein signal transduction
peptidyl-tyrosine phosphorylation
positive regulation of Arp2/3 complex-mediated actin nucleation
DNA binding
protein binding
cytoskeletal adaptor activity
SH3 domain binding
kinase binding
ubiquitin protein ligase binding
protein complex binding
Rac GTPase binding
proline-rich region binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0