Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other R3HCC1L SIRNA Assays ›
Gene Symbol:
R3HCC1L
Gene Name:
R3H domain and coiled-coil containing 1 like
Gene Aliases:
C10orf28, GIDRP86, GIDRP88, PSORT
Chromosome Location:
Chr.10: 98134608 - 98244897 on Build GRCh38
UniGene ID:
Hs.419800
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
241541
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: R3HCC1L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256619.1
NP_001243548.1
11
3436
NM_001256620.1
NP_001243549.1
9
3343
NM_001256621.1
NP_001243550.1
8
1540
NM_014472.4
NP_055287.4
9
3339
NM_138469.2
NP_612478.2
10
3436
XM_011539639.1
XP_011537941.1
11
3693
XM_011539640.1
XP_011537942.1
12
3498
XM_011539641.1
XP_011537943.1
11
3450
XM_011539642.1
XP_011537944.1
12
3790
XM_011539643.1
XP_011537945.1
10
3377
XM_011539644.1
XP_011537946.1
10
3381
XM_011539645.2
XP_011537947.1
10
3624
XM_011539646.1
XP_011537948.1
9
3256
XM_011539647.1
XP_011537949.1
9
3258
XM_011539648.1
XP_011537950.1
8
3131
XM_011539649.1
XP_011537951.1
10
3651
XM_017016074.1
XP_016871563.1
10
4589
XM_017016075.1
XP_016871564.1
7
3089
XM_017016076.1
XP_016871565.1
10
3383
XM_017016077.1
XP_016871566.1
12
3546
XM_017016078.1
XP_016871567.1
11
3468
XM_017016079.1
XP_016871568.1
11
3482
XM_017016080.1
XP_016871569.1
9
3388
XM_017016081.1
XP_016871570.1
10
3470
XM_017016082.1
XP_016871571.1
8
3262
XM_017016083.1
XP_016871572.1
9
3278
GenBank mRNA
AF525304.1
-
11
3389
AK058187.1
-
5
1130
AK094479.1
-
6
1850
BC009231.2
-
5
1265
BC047908.1
-
8
1486
Back To Top
More Information
Gene Ontology Categories:
Process(es)
nucleotide binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0