Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286692.1 | NP_001273621.1 | 28 | 4074 | ||
NM_001286693.1 | NP_001273622.1 | 26 | 3969 | |||
NM_015668.4 | NP_056483.3 | 28 | 4110 | |||
XM_005250856.2 | XP_005250913.1 | 27 | 3797 | |||
XM_005250857.2 | XP_005250914.1 | 27 | 3797 | |||
XM_005250858.2 | XP_005250915.1 | 27 | 3794 | |||
XM_006716540.2 | XP_006716603.1 | 28 | 4224 | |||
XM_011516957.1 | XP_011515259.1 | 29 | 4405 | |||
XM_011516958.1 | XP_011515260.1 | 28 | 4303 | |||
XM_017013309.1 | XP_016868798.1 | 26 | 3638 | |||
XM_017013310.1 | XP_016868799.1 | 26 | 3635 | |||
XM_017013311.1 | XP_016868800.1 | 21 | 2653 | |||
GenBank mRNA | AK097399.1 | - | 17 | 2143 | ||
AL117544.1 | - | 9 | 934 | |||
AY009106.1 | - | 28 | 3903 | |||
BC036665.1 | - | 28 | 3961 | 5268331 | ||
BC092411.1 | - | 28 | 4107 | 30386231 |