Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATPIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016311.4 | NP_057395.1 | 1, 2 | 185 | ||
NM_178190.2 | NP_835497.1 | 1, 2 | 185 | |||
NM_178191.2 | NP_835498.1 | 1, 2 | 185 | |||
GenBank mRNA | AB029042.1 | - | 1, 2 | 113 | ||
AF114836.1 | - | 271 | ||||
AK000934.1 | - | 1, 2 | 122 | |||
AK316600.1 | - | 1, 2 | 172 | |||
AL050386.1 | - | 1, 2 | 115 | |||
AL583540.3 | - | 1, 2 | 108 | |||
BC004955.1 | - | 1, 2 | 68 | 3625379 | ||
BC009677.1 | - | 1, 2 | 143 | 3877506 | ||
BG036978.1 | - | 1, 2 | 157 | |||
BG337531.1 | - | 1, 2 | 141 | |||
BG714388.1 | - | 1, 2 | 178 | |||
BP271519.1 | - | 1, 2 | 181 | |||
BT009849.1 | - | 1, 2 | 92 | |||
CR457097.1 | - | 1, 2 | 92 | |||
DB030607.1 | - | 1, 2 | 185 |