Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164115.1 | NP_001157587.1 | 2 | 589 | ||
NM_001164116.1 | NP_001157588.1 | 3 | 613 | |||
NM_020356.3 | NP_065089.2 | 2 | 589 | |||
GenBank mRNA | AJ276678.1 | - | 3 | 613 | ||
AK027760.1 | - | 2 | 586 | |||
AK308057.1 | - | 2 | 590 | |||
AK314653.1 | - | 2 | 522 | |||
BC027951.1 | - | 2 | 539 | 5205865 |