Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016275.3 | NP_057359.2 | 2 | 309 | ||
GenBank mRNA | AF131856.1 | - | 2 | 264 | ||
AF195141.1 | - | 2 | 267 | |||
AK075006.1 | - | 2 | 282 | |||
AK300315.1 | - | 2 | 297 | |||
AY358095.1 | - | 2 | 273 | |||
AY358096.1 | - | 2 | 277 | |||
BC006012.1 | - | 2 | 300 | 4333083 | ||
BC008411.1 | - | 2 | 267 | 4132578 | ||
BC009556.1 | - | 2 | 241 | 3899430 | ||
BC009611.1 | - | 2 | 261 | 4093231 | ||
BC026350.1 | - | 2 | 289 | 4796883 | ||
BC036738.1 | - | 2 | 260 | 5540243 | ||
BC071699.1 | - | 2 | 252 | 5548565 | ||
CR457154.1 | - | 1 | 51 |