Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286771.2 | NP_001273700.1 | 2 | 211 | ||
NM_015574.1 | NP_056389.1 | 2 | 593 | |||
NM_032217.4 | NP_115593.3 | 2 | 593 | |||
NM_198889.2 | NP_942592.1 | 2 | 593 | |||
XM_005265671.3 | XP_005265728.1 | 2 | 937 | |||
XM_005265672.3 | XP_005265729.1 | 2 | 937 | |||
XM_005265673.3 | XP_005265730.1 | 2 | 937 | |||
XM_017008011.1 | XP_016863500.1 | 2 | 168 | |||
XM_017008012.1 | XP_016863501.1 | 2 | 168 | |||
XM_017008013.1 | XP_016863502.1 | 2 | 168 | |||
XM_017008014.1 | XP_016863503.1 | 2 | 937 | |||
XM_017008015.1 | XP_016863504.1 | 2 | 168 | |||
XM_017008016.1 | XP_016863505.1 | 2 | 937 | |||
XM_017008017.1 | XP_016863506.1 | 2 | 168 | |||
GenBank mRNA | AB014597.2 | - | 2 | 117 | ||
AF308285.1 | - | 2 | 580 | |||
AK300302.1 | - | 2 | 167 | |||
AK307581.1 | - | 2 | 593 | |||
BC004173.2 | - | 2 | 574 | |||
BC043394.1 | - | 2 | 562 | |||
BX648035.1 | - | 2 | 166 | |||
DC335632.1 | - | 2 | 211 |