Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC16B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033127.2 | NP_149118.2 | 16 | 1997 | ||
GenBank mRNA | AB063357.1 | - | 16 | 1997 | ||
AK090971.1 | - | 2 | 99 | |||
AK098627.1 | - | 5 | 617 | |||
AK314585.1 | - | 15 | 2001 | |||
EF125213.1 | - | 15 | 1907 |