Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERVK3-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054868.1 | - | 4 | 508 | ||
BC010118.2 | - | 4 | 466 | |||
BC011670.1 | - | 4 | 487 | 4129886 |