Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SREK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077199.2 | NP_001070667.1 | 7 | 1112 | ||
NM_001270492.1 | NP_001257421.1 | 8 | 1236 | |||
NM_001323527.1 | NP_001310456.1 | 8 | 1236 | |||
NM_001323529.1 | NP_001310458.1 | 7 | 1112 | |||
NM_001323533.1 | NP_001310462.1 | 7 | 1112 | |||
NM_001323534.1 | NP_001310463.1 | 8 | 1127 | |||
NM_001323535.1 | NP_001310464.1 | 7 | 1003 | |||
NM_139168.3 | NP_631907.1 | 8 | 1265 | |||
XM_011543171.2 | XP_011541473.1 | 7 | 1337 | |||
XM_017009055.1 | XP_016864544.1 | 7 | 1009 | |||
XM_017009056.1 | XP_016864545.1 | 7 | 897 | |||
GenBank mRNA | AB209694.1 | - | 1 | 1253 | ||
AF459094.1 | - | 7 | 946 | |||
AK091758.1 | - | 7 | 995 | |||
AK308287.1 | - | 8 | 1139 | |||
AL832888.1 | - | 4 | 1969 | |||
BC047322.1 | - | 7 | 981 | |||
BC067770.1 | - | 8 | 1266 | 4827589 | ||
BC112343.1 | - | 5 | 634 | 40008558 |