Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARPP21 SIRNA Assays ›
Gene Symbol:
ARPP21
Gene Name:
cAMP regulated phosphoprotein 21
Gene Aliases:
ARPP-21, R3HDM3, RCS, TARPP
Chromosome Location:
Chr.3: 35638981 - 35798933 on Build GRCh38
UniGene ID:
Hs.475902
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
24198
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARPP21
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025068.1
NP_001020239.1
5
464
NM_001025069.1
NP_001020240.1
5
473
NM_001267616.1
NP_001254545.1
5
1620
NM_001267617.1
NP_001254546.1
5
430
NM_001267618.1
NP_001254547.1
4
561
NM_001267619.1
NP_001254548.1
4
395
NM_016300.4
NP_057384.2
5
447
NM_198399.1
NP_938409.1
4
455
XM_005264811.2
XP_005264868.1
5
476
XM_006712943.2
XP_006713006.1
5
476
XM_006712944.2
XP_006713007.1
5
476
XM_011533298.2
XP_011531600.1
5
476
XM_011533299.2
XP_011531601.1
5
1020
XM_011533300.2
XP_011531602.1
5
639
XM_011533301.2
XP_011531603.1
5
470
XM_011533302.2
XP_011531604.1
6
562
XM_011533303.2
XP_011531605.1
6
1112
XM_017005574.1
XP_016861063.1
5
566
XM_017005575.1
XP_016861064.1
6
12199
XM_017005576.1
XP_016861065.1
5
1020
XM_017005577.1
XP_016861066.1
5
470
XM_017005578.1
XP_016861067.1
5
446
XM_017005579.1
XP_016861068.1
5
1020
XM_017005580.1
XP_016861069.1
6
1112
XM_017005581.1
XP_016861070.1
6
12199
XM_017005582.1
XP_016861071.1
5
1020
XM_017005583.1
XP_016861072.1
5
470
XM_017005584.1
XP_016861073.1
5
1020
XM_017005585.1
XP_016861074.1
5
1020
XM_017005586.1
XP_016861075.1
5
476
XM_017005587.1
XP_016861076.1
5
639
XM_017005588.1
XP_016861077.1
6
1112
XM_017005589.1
XP_016861078.1
6
562
XM_017005590.1
XP_016861079.1
6
12199
XM_017005591.1
XP_016861080.1
5
1020
XM_017005592.1
XP_016861081.1
5
1020
XM_017005593.1
XP_016861082.1
5
566
XM_017005594.1
XP_016861083.1
5
470
XM_017005595.1
XP_016861084.1
6
1112
XM_017005596.1
XP_016861085.1
5
1020
XM_017005597.1
XP_016861086.1
5
1020
XM_017005598.1
XP_016861087.1
6
1112
XM_017005599.1
XP_016861088.1
5
1020
XM_017005600.1
XP_016861089.1
5
639
XM_017005601.1
XP_016861090.1
5
470
XM_017005602.1
XP_016861091.1
5
1020
XM_017005603.1
XP_016861092.1
5
639
XM_017005604.1
XP_016861093.1
5
470
XM_017005605.1
XP_016861094.1
6
1112
XM_017005606.1
XP_016861095.1
6
562
XM_017005607.1
XP_016861096.1
5
1020
XM_017005608.1
XP_016861097.1
5
1020
XM_017005609.1
XP_016861098.1
6
1112
XM_017005610.1
XP_016861099.1
5
1020
XM_017005611.1
XP_016861100.1
5
1020
XM_017005612.1
XP_016861101.1
5
1020
GenBank mRNA
AF112220.1
-
4
420
AK055064.1
-
5
473
AK289649.1
-
4
410
AK289868.1
-
5
467
AK294484.1
-
4
408
AV718064.1
-
5
464
AV718166.1
-
5
473
BC017805.1
-
4
455
BC036399.1
-
5
435
5263894
BC041385.1
-
5
1208
BC051828.1
-
5
447
6012704
BC066651.1
-
3
974
BX537647.1
-
1
577
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
cellular response to heat
molecular_function
nucleic acid binding
calmodulin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0