Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083961.1 | NP_001077430.1 | 23 | 2912 | ||
NM_173636.4 | NP_775907.4 | 23 | 2912 | |||
XM_005258809.2 | XP_005258866.1 | 23 | 2912 | |||
XM_011526837.1 | XP_011525139.1 | 23 | 2897 | |||
XM_011526838.1 | XP_011525140.1 | 23 | 2912 | |||
XM_011526839.1 | XP_011525141.1 | 21 | 2561 | |||
XM_011526840.2 | XP_011525142.1 | 16 | 1907 | |||
XM_011526841.2 | XP_011525143.1 | 14 | 1528 | |||
XM_011526842.1 | XP_011525144.1 | 13 | 1360 | |||
XM_011526843.1 | XP_011525145.1 | 6 | 687 | |||
XM_011526844.2 | XP_011525146.1 | 6 | 684 | |||
XM_017026665.1 | XP_016882154.1 | 23 | 2912 | |||
GenBank mRNA | AL133651.1 | - | 2 | 229 | ||
BC017261.2 | - | 5 | 621 | |||
BC058939.1 | - | 23 | 2856 | |||
BX647726.1 | - | 24 | 2973 | |||
KU178897.1 | - | 2 | 157 | |||
KU178898.1 | - | 2 | 157 | |||
KU178899.1 | - | 2 | 157 |