Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPRG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182752.3 | NP_877429.2 | 5 | 2264 | ||
GenBank mRNA | AK025409.1 | - | 1 | 674 | ||
AK025464.1 | - | 4 | 1960 | |||
AK096427.1 | - | 5 | 2218 | |||
BC019034.2 | - | 4 | 2006 | 4576259 | ||
BX115868.1 | - | 132 |