Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBEAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114132.1 | NP_001107604.1 | 54 | 8258 | ||
XM_005246787.3 | XP_005246844.1 | 54 | 8284 | |||
XM_005246788.2 | XP_005246845.1 | 33 | 4760 | |||
XM_006712698.3 | XP_006712761.1 | 55 | 8371 | |||
XM_006712699.3 | XP_006712762.1 | 55 | 8371 | |||
XM_011511658.2 | XP_011509960.1 | 55 | 8326 | |||
XM_011511659.2 | XP_011509961.1 | 55 | 8383 | |||
XM_011511660.2 | XP_011509962.1 | 51 | 7705 | |||
XM_011511662.2 | XP_011509964.1 | 47 | 7040 | |||
XM_017004728.1 | XP_016860217.1 | 42 | 6438 | |||
XM_017004729.1 | XP_016860218.1 | 30 | 4452 | |||
GenBank mRNA | AB053319.1 | - | 4 | 509 | ||
AK127772.1 | - | 27 | 3856 | |||
AK128636.1 | - | 24 | 2985 | |||
AY172970.1 | - | 25 | 3164 | |||
BX647133.1 | - | 18 | 2085 |