Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBEAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114132.1 | NP_001107604.1 | 4 | 496 | ||
XM_005246787.3 | XP_005246844.1 | 4 | 522 | |||
XM_006712698.3 | XP_006712761.1 | 4 | 522 | |||
XM_006712699.3 | XP_006712762.1 | 4 | 522 | |||
XM_011511658.2 | XP_011509960.1 | 4 | 477 | |||
XM_011511659.2 | XP_011509961.1 | 4 | 534 | |||
XM_011511663.2 | XP_011509965.1 | 4 | 512 | |||
GenBank mRNA | AB053318.1 | - | 4 | 410 | ||
BC132950.1 | - | 4 | 450 | |||
BC137214.1 | - | 4 | 450 | |||
BC144532.1 | - | 4 | 450 | |||
BC144533.1 | - | 4 | 445 |