Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000858.5 | NP_000849.1 | 8 | 809 | ||
NM_001159390.1 | NP_001152862.1 | 7 | 644 | |||
NM_001159391.1 | NP_001152863.1 | 7 | 558 | |||
NM_001242839.1 | NP_001229768.1 | 7 | 584 | |||
XM_005273104.4 | XP_005273161.1 | 9 | 978 | |||
XM_017001092.1 | XP_016856581.1 | 8 | 1297 | |||
XM_017001095.1 | XP_016856584.1 | 8 | 754 | |||
XM_017001096.1 | XP_016856585.1 | 7 | 594 | |||
GenBank mRNA | AK124677.1 | - | 3356 | |||
AK303845.1 | - | 7 | 597 | |||
BC006249.2 | - | 8 | 747 | 3946293 | ||
BC007369.2 | - | 6 | 461 | |||
BC009914.2 | - | 8 | 721 | 2821563 | ||
BC091480.1 | - | 7 | 564 | 30340102 | ||
BE780558.1 | - | 7 | 531 | |||
BI771824.1 | - | 7 | 584 | |||
CR456836.1 | - | 6 | 506 | |||
CR542006.1 | - | 6 | 506 | |||
L76200.1 | - | 6 | 506 | |||
U66895.1 | - | 6 | 512 |