Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKCI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002740.5 | NP_002731.4 | 2 | 373 | ||
XM_017006860.1 | XP_016862349.1 | 2 | 366 | |||
GenBank mRNA | AB451443.1 | - | 2 | 108 | ||
AK315342.1 | - | 2 | 301 | |||
BC022016.2 | - | 2 | 368 | 4823853 | ||
L18964.1 | - | 2 | 372 | |||
L33881.1 | - | 2 | 312 |