Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STYK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018423.2 | NP_060893.2 | 11 | 2339 | ||
XM_005253417.2 | XP_005253474.1 | 10 | 2239 | |||
XM_011520736.1 | XP_011519038.1 | 12 | 2142 | |||
XM_011520737.2 | XP_011519039.1 | 11 | 2179 | |||
XM_011520738.2 | XP_011519040.1 | 11 | 2334 | |||
XM_017019579.1 | XP_016875068.1 | 11 | 2122 | |||
GenBank mRNA | AF251059.1 | - | 11 | 2339 | ||
AL353940.1 | - | 11 | 2087 |