Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PACSIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199583.2 | NP_001186512.1 | 10 | 2645 | ||
NM_020804.4 | NP_065855.1 | 10 | 2658 | |||
XM_011514541.1 | XP_011512843.1 | 10 | 2513 | |||
GenBank mRNA | AB037800.2 | - | 10 | 2615 | ||
AL834211.1 | - | 10 | 2591 | |||
BC040228.1 | - | 10 | 2630 | 5173129 |