Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135040.2 | NP_001128512.1 | 16, 17 | 2510 | ||
NM_001135041.2 | NP_001128513.1 | 14, 15 | 2233 | |||
NM_001190836.1 | NP_001177765.1 | 16, 17 | 2453 | |||
NM_001190837.1 | NP_001177766.1 | 18, 19 | 2549 | |||
NM_004082.4 | NP_004073.2 | 19, 20 | 2570 | |||
NM_023019.3 | NP_075408.1 | 14, 15 | 2233 | |||
NR_033935.1 | 16, 17 | 2453 | ||||
GenBank mRNA | AB209625.1 | - | 10, 11 | 3069 | ||
AK297286.1 | - | 16, 17 | 2205 | |||
AK314352.1 | - | 16, 17 | 2454 | |||
AK315014.1 | - | 14, 15 | 2025 | |||
BC071583.1 | - | 14, 15 | 2150 | 4817849 | ||
BX640799.1 | - | 19, 20 | 2349 | |||
BX647925.1 | - | 16, 17 | 2214 | |||
BX647972.1 | - | 16, 17 | 2233 | |||
U90445.1 | - | 7, 8 | 1028 | |||
X98801.1 | - | 17, 18 | 2207 |