Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIAPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079812.2 | NP_001073280.1 | 27 | 5141 | ||
NM_001314007.1 | NP_001300936.1 | 29 | 5221 | |||
NM_005219.4 | NP_005210.3 | 28 | 5168 | |||
XM_011537572.2 | XP_011535874.1 | 27 | 5109 | |||
XM_011537573.2 | XP_011535875.1 | 28 | 4976 | |||
GenBank mRNA | AB209482.1 | - | 28 | 5109 | ||
AF051782.1 | - | 27 | 5015 | |||
AK057994.1 | - | 4 | 1616 | |||
AK094264.1 | - | 2 | 1979 | |||
AL050086.1 | - | 1 | 199 | |||
AY007129.1 | - | 1 | 1346 | |||
DQ067453.1 | - | 27 | 5072 |