Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014470.3 | NP_055285.1 | 5 | 1280 | ||
GenBank mRNA | AK124288.1 | - | 1 | 1768 | ||
AL359573.1 | - | 1 | 513 | |||
BC026356.1 | - | 5 | 1280 | 4793442 |