Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016246.2 | NP_057330.2 | 9 | 1215 | ||
XM_005258969.3 | XP_005259026.1 | 8 | 1127 | |||
XM_005258970.3 | XP_005259027.1 | 7 | 1037 | |||
GenBank mRNA | AF126781.1 | - | 9 | 1061 | ||
AY358430.1 | - | 9 | 1054 | |||
BC006283.2 | - | 9 | 1008 | 4026397 | ||
BC006294.1 | - | 9 | 1016 | 4025809 |