Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178844.3 | NP_849172.2 | 15, 16 | 3194 | ||
NR_075083.2 | 16, 17 | 3391 | ||||
XM_017023027.1 | XP_016878516.1 | 15, 16 | 3224 | |||
XM_017023028.1 | XP_016878517.1 | 16, 17 | 3398 | |||
XM_017023029.1 | XP_016878518.1 | 17, 18 | 3479 | |||
XM_017023030.1 | XP_016878519.1 | 17, 18 | 3540 | |||
XM_017023031.1 | XP_016878520.1 | 15, 16 | 3285 | |||
XM_017023032.1 | XP_016878521.1 | 17, 18 | 4241 | |||
XM_017023033.1 | XP_016878522.1 | 17, 18 | 3418 | |||
XM_017023034.1 | XP_016878523.1 | 16, 17 | 3337 | |||
XM_017023035.1 | XP_016878524.1 | 16, 17 | 3366 | |||
XM_017023036.1 | XP_016878525.1 | 15, 16 | 3925 | |||
XM_017023037.1 | XP_016878526.1 | 16, 17 | 3119 | |||
XM_017023038.1 | XP_016878527.1 | 16, 17 | 3092 | |||
GenBank mRNA | AK074109.1 | - | 11, 12 | 1072 | ||
AK090431.1 | - | 16, 17 | 3387 | |||
AY601811.1 | - | 15, 16 | 3181 | |||
FJ889357.1 | - | 12, 13 | 2770 |