Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002983.2 | NP_002974.1 | 3 | 653 | ||
GenBank mRNA | BC071834.1 | - | 3 | 625 | 5935287 | |
BX103455.1 | - | 3 | 653 | |||
D00044.1 | - | 3 | 634 | |||
M23452.1 | - | 3 | 634 | |||
M25315.1 | - | 3 | 634 | |||
X03754.1 | - | 3 | 634 |