Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006942.1 | NP_008873.1 | 1 | 512 | ||
GenBank mRNA | AB006867.1 | - | 1 | 512 | ||
AK301687.1 | - | 1 | 433 | |||
BC000985.2 | - | 1 | 416 | 3447385 | ||
BC072003.1 | - | 2 | 166 | 4653140 |