Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARVELD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017967.3 | NP_001017967.2 | 1 | 389 | ||
NM_052858.5 | NP_443090.4 | 1 | 389 | |||
XM_011523449.2 | XP_011521751.1 | 1 | 405 | |||
GenBank mRNA | AK056842.1 | - | 1 | 355 | ||
AK292185.1 | - | 1 | 355 | |||
BC005052.1 | - | 1 | 381 | |||
BC011893.2 | - | 1 | 356 | 3942185 | ||
BC013376.2 | - | 1 | 356 | 3946889 | ||
BM742281.1 | - | 1 | 375 | |||
BP320419.1 | - | 1 | 390 |