Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007124.2 | NP_009055.2 | 2 | 207 | ||
XM_005267127.4 | XP_005267184.1 | 2 | 542 | |||
XM_005267130.2 | XP_005267187.1 | 3 | 425 | |||
XM_005267133.2 | XP_005267190.1 | 2 | 1225 | |||
XM_011536101.2 | XP_011534403.1 | 4 | 886 | |||
XM_011536102.2 | XP_011534404.1 | 4 | 427 | |||
XM_011536106.2 | XP_011534408.1 | 3 | 1074 | |||
XM_011536107.2 | XP_011534409.1 | 3 | 450 | |||
XM_017011243.1 | XP_016866732.1 | 4 | 427 | |||
XM_017011244.1 | XP_016866733.1 | 4 | 427 | |||
XM_017011245.1 | XP_016866734.1 | 4 | 427 | |||
GenBank mRNA | AK292506.1 | - | 3 | 425 | ||
DB079517.1 | - | 3 | 425 | |||
X69086.1 | - | 2 | 115 |