Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZYG11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024646.2 | NP_078922.1 | 9 | 1771 | ||
XM_006710898.3 | XP_006710961.1 | 9 | 1658 | |||
XM_017002336.1 | XP_016857825.1 | 9 | 1776 | |||
GenBank mRNA | AB051517.1 | - | 9 | 1721 | ||
AK023518.1 | - | 2 | 139 | |||
AK056645.1 | - | 7 | 715 | |||
AK095643.1 | - | 8 | 1411 | |||
BC029832.1 | - | 7 | 785 | |||
BX647643.1 | - | 7 | 944 | |||
DQ646393.1 | - | 9 | 1790 |