Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000618.4 | NP_000609.1 | 1 | 302 | ||
NM_001111283.2 | NP_001104753.1 | 1 | 302 | |||
XM_017019260.1 | XP_016874749.1 | 1 | 331 | |||
GenBank mRNA | AB209184.1 | - | 1 | 253 | ||
AK312231.1 | - | 1 | 201 | |||
AL603526.1 | - | 1 | 269 | |||
AL710591.1 | - | 1 | 303 | |||
BC148266.1 | - | 1 | 60 | 8860206 | ||
BC152321.1 | - | 1 | 183 | 40129266 | ||
CR541861.1 | - | 1 | 38 | |||
M11568.1 | - | 1 | 220 | |||
M27544.1 | - | 1 | 186 | |||
M29644.1 | - | 1 | 49 | |||
X00173.1 | - | 1 | 49 | |||
X56773.1 | - | 1 | 38 | |||
X56774.1 | - | 1 | 38 | |||
X57025.1 | - | 1 | 204 |