Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CITED4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133467.2 | NP_597724.1 | 1 | 299 | ||
GenBank mRNA | BC035496.2 | - | 1 | 301 | 3622207 | |
BC052559.1 | - | 1 | 299 | 6250908 |