Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001890.2 | NP_001001890.1 | 3 | 2086 | ||
NM_001122607.1 | NP_001116079.1 | 3 | 2086 | |||
NM_001754.4 | NP_001745.2 | 6 | 779 | |||
XM_005261068.3 | XP_005261125.1 | 4 | 1042 | |||
XM_005261069.4 | XP_005261126.1 | 6 | 815 | |||
XM_011529766.2 | XP_011528068.1 | 6 | 890 | |||
XM_011529767.2 | XP_011528069.1 | 5 | 776 | |||
XM_011529768.2 | XP_011528070.1 | 5 | 776 | |||
XM_011529770.2 | XP_011528072.1 | 6 | 815 | |||
XM_017028487.1 | XP_016883976.1 | 4 | 584 | |||
GenBank mRNA | AK226159.1 | - | 5 | 776 | ||
AK310587.1 | - | 6 | 768 | |||
BC110828.1 | - | 5 | 674 | |||
BC136380.1 | - | 6 | 736 | 9020367 | ||
BC136381.1 | - | 6 | 735 | 9020368 | ||
BC144053.1 | - | 6 | 735 | 9052573 | ||
D10570.1 | - | 3 | 1276 | |||
D43967.1 | - | 3 | 2086 | |||
D43968.1 | - | 3 | 2086 | |||
D43969.1 | - | 5 | 1033 | |||
D89788.1 | - | 4 | 476 | |||
D89789.1 | - | 4 | 476 | |||
D89790.1 | - | 4 | 476 | |||
L34598.1 | - | 3 | 1186 | |||
S60998.1 | - | 3 | 516 | |||
S76345.1 | - | 2 | 101 | |||
S76346.1 | - | 2 | 101 | |||
U19601.1 | - | 5 | 589 | |||
X79549.1 | - | 3 | 2076 |
Gene Symbol: RUNX1T1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | Z35296.1 | - | 93 |