Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHPN1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026785.1 | 1 | 388 | |||
GenBank mRNA | AK026503.1 | - | 1 | 299 | ||
BC000203.1 | - | 1 | 301 | |||
BI829214.1 | - | 1 | 66 |