Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLJ40288 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046323.1 | 4, 5 | 661 | |||
GenBank mRNA | AK097607.1 | - | 4, 5 | 661 |