Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP3R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147180.3 | NP_671709.1 | 1 | 565 | ||
GenBank mRNA | AF085237.1 | - | 1 | 485 | ||
AF145026.1 | - | 1 | 485 | |||
AF400667.1 | - | 1 | 539 | |||
AK057524.1 | - | 1 | 524 | |||
BC030595.2 | - | 1 | 565 | 4827322 | ||
BC066299.1 | - | 1 | 506 | 5298489 |