Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025109.1 | NP_001020280.1 | 5 | 917 | ||
NM_001773.2 | NP_001764.1 | 5 | 917 | |||
GenBank mRNA | AF202879.1 | - | 3 | 162 | ||
AF523361.1 | - | 5 | 730 | |||
AF538601.1 | - | 4 | 568 | |||
AK291529.1 | - | 5 | 670 | |||
AK293030.1 | - | 5 | 694 | |||
AK294381.1 | - | 4 | 441 | |||
AK300835.1 | - | 6 | 734 | |||
BC039146.1 | - | 5 | 667 | 4746591 | ||
BX538076.1 | - | 2 | 845 | |||
M81104.1 | - | 5 | 916 | |||
S53910.1 | - | 5 | 749 | |||
S53911.1 | - | 5 | 749 |