Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KANK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320269.1 | NP_001307198.1 | 9 | 2121 | ||
NM_181712.4 | NP_859063.3 | 10 | 4003 | |||
XM_011540843.2 | XP_011539145.1 | 10 | 3888 | |||
XM_011540845.2 | XP_011539147.1 | 10 | 3882 | |||
XM_017000483.1 | XP_016855972.1 | 9 | 3752 | |||
XM_017000484.1 | XP_016855973.1 | 10 | 2257 | |||
XM_017000485.1 | XP_016855974.1 | 8 | 2189 | |||
GenBank mRNA | AB284127.1 | - | 10 | 4003 | ||
AK091941.1 | - | 9 | 2007 | |||
AK096259.1 | - | 4 | 1387 | |||
AK127461.1 | - | 7 | 2929 | |||
AL832644.2 | - | 8 | 2755 | |||
BC041418.1 | - | 8 | 2223 | |||
BC060866.1 | - | 10 | 4003 | 30344114 |