Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135186.1 | NP_001128658.1 | 8 | 1457 | ||
NM_016428.2 | NP_057512.1 | 8 | 1475 | |||
XM_005257429.2 | XP_005257486.1 | 8 | 1141 | |||
XM_011524873.1 | XP_011523175.1 | 8 | 1138 | |||
XM_017024721.1 | XP_016880210.1 | 8 | 1117 | |||
GenBank mRNA | AB037886.1 | - | 8 | 1475 | ||
AL136709.1 | - | 8 | 1276 | |||
BC007780.2 | - | 8 | 981 | 4309524 |