Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002711.3 | NP_002702.2 | 1 | 763 | ||
XM_005250473.3 | XP_005250530.1 | 2 | 309 | |||
GenBank mRNA | AF024579.1 | - | 2 | 211 | ||
BC056666.1 | - | 1 | 766 | |||
BC070313.1 | - | 1 | 745 | |||
BC126451.1 | - | 1 | 801 | 8992167 | ||
BC126453.1 | - | 1 | 801 | 8992169 | ||
X78578.1 | - | 1 | 730 |